missing translation for 'onlineSavingsMsg'
Learn More

Boric Acid BAKER ANALYZED Reagent, 500g

Brand:  JT Baker 0055.0500

60.80 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.



Boric Acid BAKER ANALYZED Reagent, 500g

Code : NEW

Additional Details : CAS Number : 10043-35-3 Weight : 0.50000kg


Product Code. 10000410

  • £64.00 / 500g
Estimated Shipment: 11-10-2024
to see stock.

Description

Description

Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Boric Acid BAKER ANALYZED Reagent, 500g

Signal Word
  • Danger

Hazard Category
  • Reproductive toxicity Category 1B

Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : ortho-Boric acid

SDS
Documents

Documents

Certificates
Ofertas especiales

Ofertas especiales