Learn More
Boscalid, Fluka™
Analytical standard, reference material
Brand: Honeywell Fluka R1667-100MG
Additional Details : CAS Number : 188425-85-6 Weight : 0.00010kg Transport : UN number : 3077 Chem class : 9 Pack group : III
Description
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
- Expert Technical Support
Specifications
Boscalid | |
188425-85-6 | |
White | |
343.21 | |
5.5 (10 g/L at 23°C) | |
Decomposes on heating. |
Glass Bottle | |
100 mg | |
2-Chloro-N-(4'-chlorobiphenyl-2- yl)-nicotinamide | |
Analytical Standard | |
148°C | |
Environmental Analysis |