missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Bossheads

For securely keeping clamps in place on laboratory scaffolding. R&L Enterprises™ Bossheads come with offset jaws for greater strength.

Brand:  Fisherbrand™ 2005/1

Product Code. 11745386

  • £15.00 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Aluminum alloy with clamping screws
  • For use with rods up to 16mm in diameter
Specifications

Specifications

1 Ea.
Blue Plastic headed screw
16 mm
Bosshead Clamp
For Rods up to 16mm dia.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.