missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Usbeck Laborgeraete™ Bosshead, Stainless Steel
Click to view available options
Material:
Stainless Steel
Unit Size:
Each
Description
- Electrolytically Polished
- For rods up to 16 mm diameter
- Screws: T-Screws
- Material: Stainless Steel
- Weight: 119 g
Specifications
Specifications
| Material | Stainless Steel |
| Size (Metric) Grip | 16 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction