Learn More
R&D Systems™ MgATP Solution
General Ubiquitin Proteasome Pathway (UPP) Products
Brand: R&D Systems™ B-20
Additional Details : Weight : 0.00970kg
Description
Pre-coupled Mg-ATP is an ideal energy source for semi-purified conjugation/degradation reactions. Supplied as a pH adjusted solution with no added buffering reagents.Stock: 0.1M Mg2+-ATP, pH 7.0
Specifications
Store the unopened product at -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |