missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Socorex™ Calibrex™ Universal 520 Bottle-top Dispensers
Constructed with chemically inert contact components for broad chemical resistance, these bottle-top dispensers are universally adapted for most laboratory reagents. Socorex™ Calibrex™ Universal 520 Bottle-top Dispensers are easy to use and maintain.
Brand: Socorex™ 520.010
This item is not returnable.
View return policy
Description
- Adjustable volume dispenser.
- Chemically resistant.
- Smooth, effortless activation.
- Base thread fits 32mm bottle neck diameter.
- Comes with 3 adapters (PP material) for 28, 40 and 45mm bottle neck diameters.
- Color coding identification.
- QR coded chemical compatibility
- Fast, reliable and friendly volume setting:
- Fast and precise single-hand adjustment.
- Efficient click-stop mechanism prevents unwanted alteration.
- Large display is easy to read; the window adjusts to desired side of instrument body.
- Each step on the cylindrical cam is pre-calibrated and corresponds to one division on the volume graduation.
- Instrument rotates 360° for adequate positioning on top of the bottle.
- Air inlet can be enlarged to receive a Luer membrane filter if airborne contamination is a concern.
- Factory calibrated and easily recalibrated in the laboratory.
- Disassembly/reassembly without tools.
- Autoclavable at 121°C / 250°F.
Specifications
<±1.5% - <±0.6% | |
Autoclavable | |
0.25 mL |
Bottle-Calibrex | |
1 to 10 mL | |
<0.5% - <0.1% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Socorex™ Calibrex™ Universal 520 Bottle-top Dispensers > Volume: 1-10mL
Spot an opportunity for improvement?Share a Content Correction