missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Bottle Top Dispenser (BTD) PTFE Plunger
PTFE plunger for BTD and dispensing reagents
£31.56 - £82.07
Specifications
For Use With (Application) | Dispensing Organic and Inorganic Liquids |
---|---|
Includes | Four adapters |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
16802110
|
Corning™
6861 |
2.5 mL |
£31.56
Each |
Estimated Shipment: 12-12-2024
Log in to see stock. |
|||||
16812110
|
Corning™
6862 |
5 mL |
£34.72
Each |
Estimated Shipment: 12-12-2024
Log in to see stock. |
|||||
16822110
|
Corning™
6863 |
10 mL |
£41.04
Each |
Estimated Shipment: 12-12-2024
Log in to see stock. |
|||||
16832110
|
Corning™
6864 |
25 mL |
£51.56
Each |
Estimated Shipment: 12-12-2024
Log in to see stock. |
|||||
16842110
|
Corning™
6865 |
50 mL |
£65.24
Each |
Estimated Shipment: 12-12-2024
Log in to see stock. |
|||||
16852110
|
Corning™
6866 |
100 mL |
£82.07
Each |
Estimated Shipment: 12-12-2024
Log in to see stock. |
|||||
Description
- Specialized airpurging and closed circuit system ensures no loss of reagent during dispensing, as well as when the device is not in use
- Comes with four adapters with different diameters for universal use with multiple bottle sizes
- Telescopic suction tube can be adjusted smoothly to different bottle heights
- Calibration tool allows easy in-lab calibration with built-in recalibration mechanism to ensure the highest reproducibility during the dispenser’s long life cycle
- Every dispenser is individually tested and delivered with a certificate of performance
- For dispensing organic and inorganic liquids
Specifications
Dispensing Organic and Inorganic Liquids | |
Four adapters |