missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Bottle Top Dispensers
Corning™ Bottle Top Dispensers are volumetric instruments designed for convenient and safe dispensing of liquids with no loss of reagent.
Brand: Corning™ 6842
361.63 GBP valid until 2025-04-13
Use promo code "24122" to get your promotional price.
This item is not returnable.
View return policy
Description
- Accurate and reproducible volume setting
- 360° rotatable valve block for convenient dispensing with tough bottle connections
- Convenient 3-position discharge tube for dispensing, sample recovery, and emptying
- Unique drip-free system to increase safety at work
- Easy disassembly for cleaning
- Easy in-lab calibration with built-in recalibration mechanism

Specifications
±0.06 mL | |
ISO 8655 | |
2.2 g/cm3 | |
1 to 10 mL | |
Glass and Plastic Bottles | |
0.2 mL | |
Non-sterile | |
1°C to 40°C | |
≤ 500 mm2/sec |
Bottletop Dispenser | |
Gray, Orange | |
Polypropylene, PTFE | |
Quick and precise dispensing of acids, bases and solvents | |
Calibration Tool, Thread Adaptors Set (4 sizes), Telescopic Suction Tube, Instruction Manual, Certificate of Performance | |
≤ 0.010 mL | |
33.8°F to 104°F | |
≤ 500 mbar | |
3 Years |
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
New Offer Save Up to 92%
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Corning™ Bottle Top Dispensers > 1 to 10 mL
Spot an opportunity for improvement?Share a Content Correction