missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Bottle Top Dispensers

Product Code. 16822090 Shop All Corning Products
Click to view available options
Volume (Metric):
0.25 to 2.5 mL
0.5 to 5 mL
1 to 10 mL
10 to 100 mL
2.5 to 25 mL
5 to 50 mL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 16822090

Brand: Corning™ 6843

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Corning™ Bottle Top Dispensers are volumetric instruments designed for convenient and safe dispensing of liquids with no loss of reagent.

  • Accurate and reproducible volume setting
  • 360° rotatable valve block for convenient dispensing with tough bottle connections
  • Convenient 3-position discharge tube for dispensing, sample recovery, and emptying
  • Unique drip-free system to increase safety at work
  • Easy disassembly for cleaning
  • Easy in-lab calibration with built-in recalibration mechanism
TRUSTED_SUSTAINABILITY

Specifications

Accuracy ±0.15 mL
Product Type Bottletop Dispenser
Certifications/Compliance ISO 8655
Color Gray, Orange
Density 2.2 g/cm3
Material Polypropylene, PTFE
Volume (Metric) 2.5 to 25 mL
For Use With (Application) Quick and precise dispensing of acids, bases and solvents
For Use With (Equipment) Glass and Plastic Bottles
Includes Calibration Tool, Thread Adaptors Set (4 sizes), Telescopic Suction Tube, Instruction Manual, Certificate of Performance
Increments 0.5 mL
Precision ≤ 0.025 mL
Sterility Non-sterile
Temperature (English) 33.8°F to 104°F
Temperature (Metric) 1°C to 40°C
Vapor Pressure ≤ 500 mbar
Viscosity Rating ≤ 500 mm2/sec
Warranty 3 Years
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.