missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Bottle Top Dispensers

Corning™ Bottle Top Dispensers are volumetric instruments designed for convenient and safe dispensing of liquids with no loss of reagent.

Brand:  Corning™ 6844

518.40 GBP valid until 2025-04-13
Use promo code "24122" to get your promotional price.


 View more versions of this product

Product Code. 16832090

  • £545.68 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Accurate and reproducible volume setting
  • 360° rotatable valve block for convenient dispensing with tough bottle connections
  • Convenient 3-position discharge tube for dispensing, sample recovery, and emptying
  • Unique drip-free system to increase safety at work
  • Easy disassembly for cleaning
  • Easy in-lab calibration with built-in recalibration mechanism
TRUSTED_SUSTAINABILITY
Specifications

Specifications

±0.3 mL
ISO 8655
2.2 g/cm3
5 to 50 mL
Glass and Plastic Bottles
1 mL
Non-sterile
1°C to 40°C
≤ 500 mm2/sec
Bottletop Dispenser
Gray, Orange
Polypropylene, PTFE
Quick and precise dispensing of acids, bases and solvents
Calibration Tool, Thread Adaptors Set (4 sizes), Telescopic Suction Tube, Instruction Manual, Certificate of Performance
≤ 0.050 mL
33.8°F to 104°F
≤ 500 mbar
3 Years
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ Bottle Top Dispensers > 5 to 50 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.