missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Bottle Top Dispensers (BTD) Outer Housing
Outer housing for BTD and dispensing reagents
£38.20 - £92.77
Specifications
For Use With (Application) | Dispensing Organic and Inorganic Liquids |
---|---|
Includes | Four adapters |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
16852090
|
Corning™
6846 |
2.5 mL |
£38.20
Each |
Estimated Shipment: 02-05-2025
Log in to see stock. |
|||||
16862090
|
Corning™
6847 |
5 mL |
£38.20
Each |
Estimated Shipment: 02-05-2025
Log in to see stock. |
|||||
16872090
|
Corning™
6848 |
25 mL |
£69.86
Each |
Estimated Shipment: 02-05-2025
Log in to see stock. |
|||||
16882090
|
Corning™
6849 |
50 mL |
£69.86
Each |
Estimated Shipment: 02-05-2025
Log in to see stock. |
|||||
16892090
|
Corning™
6850 |
100 mL |
£92.77
Each |
Estimated Shipment: 02-05-2025
Log in to see stock. |
|||||
Descrizione
- Specialized airpurging and closed circuit system ensures no loss of reagent during dispensing, as well as when the device is not in use
- Comes with four adapters with different diameters for universal use with multiple bottle sizes
- Telescopic suction tube can be adjusted smoothly to different bottle heights
- Calibration tool allows easy in-lab calibration with built-in recalibration mechanism to ensure the highest reproducibility during the dispenser’s long life cycle
- Every dispenser is individually tested and delivered with a certificate of performance
- For dispensing organic and inorganic liquids
Specifica
Dispensing Organic and Inorganic Liquids | |
Four adapters |