Learn More
Sartorius Bottled Media for Sartorius Membrane Filtration Units
Lysine agar media for wild yeasts determination
Brand: Sartorius 14148----------A
Code : NEW
Additional Details : Weight : 0.50000kg
Description
The traditional culture media for microorganisms is agar media. This can be used for the membrane filtration method or for direct incubation. Agar media in bottles are the cost-effective alternative for casting plates.
- 250mL bottles, 4 bottles per box
- Economical, provides time and cost savings
- According to international formulas
- Consistent quality
Specifications
Liquid | |
VLB-S7-S | |
Membrane filtration method or for direct incubation, for determination of LactoBacilli and Pediococci and other beer spoiling organisms | |
250 mL |
Germany | |
250 mL | |
Bottle |
Safety and Handling
- X4 250ML MEDIA VLB-S7-S ST
Signal Word
- Danger
Hazard Category
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P102-Keep out of reach of children.
- P103-Read label before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : nitrocellulose