missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Bovine anti-Chicken IgY (H+L) Secondary Antibody, Biotin
Bovine Polyclonal Secondary Antibody
Brand: Invitrogen™ SA19505
This item is not returnable.
View return policy
Description
This antibody reacts with the heavy and light chains of chicken IgY antibody shows minimum cross-reactivity with IgG of mammalian species. SA1-9505 immunogen is immunoglobulin isolated from chicken serum.
Anti-Chicken secondary antibodies are affinity-purified antibodies with well-characterized specificity for the chicken immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
| Chicken IgY (H+L) | |
| Polyclonal | |
| Biotin | |
| Bovine | |
| Affinity chromatography | |
| RUO | |
| Chicken | |
| Secondary Antibody | |
| IgG |
| Western Blot, Immunocytochemistry, Immunohistochemistry, ELISA | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide; pH 7.6 | |
| Immunoglobulin isolated from chicken serum. | |
| 500 μg | |
| Secondary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Invitrogen™ Bovine anti-Chicken IgY (H+L) Secondary Antibody, Biotin > 500 μg; Biotin
Spot an opportunity for improvement?Share a Content Correction


