Learn More
Invitrogen™ Bovine anti-Human IgG (H+L) Secondary Antibody
Bovine Polyclonal Secondary Antibody
Brand: Invitrogen™ SA19501
Description
SA1-9501 reacts with the heavy and light chains of human IgG, and shows some cross-reactivity with IgG of other species. SA1-9501 has been successfully used in ELISA and Western blot applications. The SA1-9501 immunogen is purified human IgG.
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
| Human IgG (H+L) | |
| Polyclonal | |
| Unconjugated | |
| Bovine | |
| Affinity chromatography | |
| RUO | |
| Human | |
| Secondary Antibody | |
| IgG |
| ELISA, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Purified human IgG. | |
| 500 μg | |
| Secondary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.