Learn More
Tuttnauer Bowie and Dick Test
The Bowie and Dick Test is a special program to test steam penetration to porous and hollow loads. The cycle includes 3.5 minutes sterilization after pre-vacuum phase.
Brand:  Tuttnauer ADD222-0501
Additional Details : Weight : 0.01100kg
Product Code. 17819558
  • £589.36 / Each
Estimated Shipment: 19-08-2024
to see stock.
Description
Description
  • Special program to test steam penetration to porous and hollow loads.
  • The cycle includes 3.5 minutes sterilization after pre-vacuum phase.
Specifications
Specifications
Bowie and Dick Test
Non-sterile