missing translation for 'onlineSavingsMsg'
Learn More

SCAT Box for Politainer (Foldable Waste Container)

Product Code. 10282420
missing translation for 'orderingAttributeHoverText'
Capacity (Metric):
5 L
10 L
20 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Outer carton for the SCAT Politainer - also known as folding canisters.

SCAT Politainers are an ideal way of not having to dispense with safe storage even in limited spaces

Specifications

Capacity (Metric) 5 L
Color White (Translucent)
Dimensions (L x W x H) 190 x 189 x 186 mm
For Use With (Application) Waste Disposal System
For Use With (Equipment) Politainer
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'