missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BP 554 maleate, Tocris Bioscience™
Selective 5-HT1A agonist
Chemical Identifiers
CAS | 1221401-95-1 |
---|---|
Molecular Formula | C24H28N2O7 |
Molecular Weight (g/mol) | 456.495 |
InChI Key | NBCXNOHQTALBRA-BTJKTKAUSA-N |
Synonym | bp 554 maleate, 1-3-3,4-methylenedioxyphenoxy propyl-4-phenyl-piperazine maleate, 1-3-3,4-methylenedioxyphenoxy propyl-4-phenylpiperazine maleate, 1-3-benzo d 1,3 dioxol-5-yloxy propyl-4-phenylpiperazine maleate, 1-3-2h-1,3-benzodioxol-5-yloxy propyl-4-phenylpiperazine; maleic acid |
PubChem CID | 56972150 |
IUPAC Name | 1-[3-(1,3-benzodioxol-5-yloxy)propyl]-4-phenylpiperazine;(Z)-but-2-enedioic acid |
SMILES | C1CN(CCN1CCCOC2=CC3=C(C=C2)OCO3)C4=CC=CC=C4.C(=CC(=O)O)C(=O)O |
Description
Selective 5-HT1A agonist.Chemical Identifiers
1221401-95-1 | |
456.495 | |
bp 554 maleate, 1-3-3,4-methylenedioxyphenoxy propyl-4-phenyl-piperazine maleate, 1-3-3,4-methylenedioxyphenoxy propyl-4-phenylpiperazine maleate, 1-3-benzo d 1,3 dioxol-5-yloxy propyl-4-phenylpiperazine maleate, 1-3-2h-1,3-benzodioxol-5-yloxy propyl-4-phenylpiperazine; maleic acid | |
1-[3-(1,3-benzodioxol-5-yloxy)propyl]-4-phenylpiperazine;(Z)-but-2-enedioic acid |
C24H28N2O7 | |
NBCXNOHQTALBRA-BTJKTKAUSA-N | |
56972150 | |
C1CN(CCN1CCCOC2=CC3=C(C=C2)OCO3)C4=CC=CC=C4.C(=CC(=O)O)C(=O)O |
Specifications
1221401-95-1 | |
bp 554 maleate, 1-3-3,4-methylenedioxyphenoxy propyl-4-phenyl-piperazine maleate, 1-3-3,4-methylenedioxyphenoxy propyl-4-phenylpiperazine maleate, 1-3-benzo d 1,3 dioxol-5-yloxy propyl-4-phenylpiperazine maleate, 1-3-2h-1,3-benzodioxol-5-yloxy propyl-4-phenylpiperazine; maleic acid | |
C1CN(CCN1CCCOC2=CC3=C(C=C2)OCO3)C4=CC=CC=C4.C(=CC(=O)O)C(=O)O | |
456.495 | |
456.48 | |
BP 554 maleate |
C24H28N2O7 | |
NBCXNOHQTALBRA-BTJKTKAUSA-N | |
1-[3-(1,3-benzodioxol-5-yloxy)propyl]-4-phenylpiperazine;(Z)-but-2-enedioic acid | |
56972150 | |
>99% |
Safety and Handling
Recommended Storage : Store at Room Temperature