Learn More
Trajan Scientific and Medical™ BP5 GC Capillary Columns
Used for a wide range of applications. Trajan Scientific and Medical BP5 GC Capillary Columns are general purpose phase columns.
Brand:  Trajan Scientific and Medical™ 054203
 View more versions of this product
Product Code. 15218663
  • £499.00 / Each
Estimated Shipment: 04-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Excellent general purpose column
  • Low bleed
  • Non-polar High temperature
  • Ideal for aromatics, pesticides, herbicides, drugs of abuse and hydrocarbons
Specifications
Specifications
0.25 mm
Drugs of Abuse Analysis, Pesticide Analysis, Herbicides Analysis, Drugs of Abuse, Hydrocarbons, GC/MS
1 μm
5% Phenyl, 95% Dimethyl Polysiloxane
BP5 GC Capillary Column
-60°C to +320/340°C
30 m
BP5