missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BPC ReadyMate™ to 2X ReadyMate Y Piece Tubing Set

Minimize prep time and maximize production for media fill trials with ready-to-use media in BioProcess Containers. Multiple connections give greater flexibility.

Brand:  Thermo Scientific™ BP0080A

145.93 GBP valid until 2024-12-31
Use promo code "23263" to get your promotional price.



Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15239397

  • £161.00 / Each
Estimated Shipment: 05-08-2024
to see stock.
Add to basket

Description

Description

Minimize prep time and maximize production for media fill trials with ready-to-use media in BioProcess Containers. Create a sterile union to allow additional aseptic or steam connections using the Thermo Scientific™ BPC ReadyMate™ to 2X ReadyMate Y Piece Tubing Set.

  • Achieve effective process simulations, with minimized risk of secondary contamination
  • Minimize preparation time
  • Satisfy regulatory compliance
  • Save time, reduce labor costs
Multiple plug-and-play connection options, such as BPC ReadyMate to 2X ReadyMate, are available for greater flexibility.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
ReadyMate™ is a trademark or registered trademark of the gE Healthcare BioSciences AB.
Specifications

Specifications

Multi
Bioprocess Contaners (prepared media)
Tubing and connectors
1 m
BPC ReadyMate to 2X ReadyMate Y Piece Tubing Set
Microbiology Media
1/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers