Learn More
Thermo Scientific™ BPC ReadyMate™ to Steam Through Tubing Set
Minimize prep time and maximize production for media fill trials with ready-to-use media in BioProcess Containers. Multiple connections give greater flexibility.
Brand: Thermo Scientific™ BP0070A
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Minimize prep time and maximize production for media fill trials with ready-to-use media in BioProcess Containers. Create a sterile union to allow additional aseptic or steam connections using the Thermo Scientific™ BPC ReadyMate™ to Steam Through Tubing Set.
- Achieve effective process simulations, with minimized risk of secondary contamination
- Minimize preparation time
- Satisfy regulatory compliance
- Save time, reduce labor costs
Specifications
Multi | |
Bioprocess Contaners (prepared media) | |
Tubing and connectors | |
1 m |
BPC ReadyMate to Steam Through Tubing Set | |
Microbiology Media | |
1/Pk. |