Learn More
Thermo Scientific Chemicals Bradford Dye Reagent, Ready-to-Use soln.
Suitable for protein concentration determination (100 to 1500μg)
Brand: Thermo Scientific Chemicals J61522.K2
113.05 GBP valid until 2024-12-31
Use promo code "21665" to get your promotional price.
Description
The Bradford Reagent is suitable for protein concentration determination (100-1500μg).Bradford dye reagent is used in the determination of protein content in cell fractions and assessing the protein concentrations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Bradford Dye Reagent | |
Ready-to-Use soln. | |
Miscible with water. | |
Liquid |
Blue | |
UN1805 | |
1 L |
Safety and Handling
- Bradford Dye Reagent
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 2
Hazard Statement
- H290-May be corrosive to metals.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- H371-May cause damage to organs.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Phosphoric acid, Methanol
RUO – Research Use Only