missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Accessories for BMP71 Label Printer
Printer case, Quick charger and Printer battery for use with BMP-71 Label Printer (printer sold separately).
Brand: Brady™ 114881
This item is not returnable.
View return policy
Description
- BMP™71 Label Printer Accessories from Brady™
- Printer Case designed to protect your printer against impacts and dust while allowing for easy mobility.
- Printer Battery offers long battery life - prints up to 3900 1 in. x 2 in. labels per charge, enables cordless printer operation so you can print labels where you need them, when you need them.
- BMP™71 Quick Charger, charges battery in one hour.
Specifications
Hard Case | |
BMP71 Label Printer | |
Hard-sided Carry Case |
Black | |
Non-sterile | |
1 Ea. |