Learn More
Brady™ B-427 Self Laminating Vinyl Bulk BMP71 Printer Labels
White/translucent label for wire and cable marking
Brand:  Brady™ 114873
 View more versions of this product
Product Code. 11791755
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Features white printable area with clear “tail” that wraps around marker
  • Resistant to abrasion, solvents, water, oil and dirt
Applications:
Data Communications, Electrical, Laboratory, Healthcare, Cable Labels
Certifications
UL listed
Specifications
Specifications
UL Recognised to UL969 Labelling and Marking Standard when printed with the Brady Series R4300 and R6200 ribbons. See UL files MH17154 for specific details. Brady customers should specify at purchase if they require UL Recognised B-427 labels. Brady sel
Permanent Acrylic
Matte
Bulk Labels
BMP™71 Printer
1000/Roll
1 x 2.5 in.
-70°C to +70°C
66.68 mm
63.50 x 25.40 mm
25.4 mm
63.5 mm
Bulk BMP71 Label Printer Labels
White, Translucent
Self-laminating Vinyl (B-427)
Communications cables, Laboratory identification, Straws, Voice/data identification
1
Rectangle
25.4 x 25.4 mm
0.102 mm
3 to 0000 AWG
1 in.
2.5 in.
19.05 mm