Learn More
Brady™ BBP™ Cutter Cleaning Tool
Use the BBP™ Cutter Cleaning Tool to scrape residue off the cutting blade while cleaning your B30 Series Label Printer.
Brand: Brady™ 142116
Code : NEW
Additional Details : Weight : 0.01130kg
Description
- Package includes Cutter Cleaning Tool only (printer sold separately)
- For use with B30 Series Label Printers, including the BBP™31 and BBP™33 Label Printers
- Clean the printer cutter regularly to keep your label printer at optimal performance levels
- Use a printer cleaning kit (not included) that includes Isopropyl Alcohol soaked soft foam swabs to wipe away any remaining residue
Specifications
Cutter Tool | |
BBP31 Sign and Label Printer | |
1 Tool | |
1 Ea. |
Blue | |
Non-sterile | |
Full ranges of labels for cable and wire ID, facility and safety ID, product ID and circuit board ID |