Learn More
Brady™ BBP™ Repositional Vinyl Film
BBP™ Repositional Vinyl Film (B-581) is a durable, flexible, high gloss material ideal for indoor marking where frequent repositioning is required.
Brand:  Brady™ 118008
172.40 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.
 View more versions of this product
Product Code. 11756465
  • £235.00 / Each
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Special aggressive adhesive stays in place but removes cleanly without leaving adhesive residue making it ideal for applications like 5S and inventory
  • Vinyl film can be thermal transfer printed
  • Average outdoor durability is 5 years
  • Service temperature of -40°F to 180°F (-40°C to 82°C)
Specifications
Specifications
B30C-2250-581-YL
Yellow
Film
Non-sterile
BBP30, BBP31, BBP33, BBP35, and BBP37 printers
1 Roll
-40°C to +82°C
100 ft.
2.25 in.
Acrylic, Repositionable, Pressure-sensitive
Gloss
Vinyl
Ideal for temporary, Removable and repositionable labels, like 5S and lean inventory label
5 Years
-40 to 180°F
0.005 in.
30.48 m