Learn More
Brady™ FreezerBondz™ Cryogenic Matte Polyester Labels for 3 in. Core Printers
Securely mark lab samples on smooth flat surfaces in challenging environments like liquid nitrogen and freezers with B-490 FreezerBondz™ Cryogenic Matte Polyester Labels.
Brand:  Brady™ 011264
220.69 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.
 View more versions of this product
Product Code. 10362273
  • £317.00 / 1 roll
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Printable labels for thermal transfer printers
  • Freezerbondz labels can be applied to frozen surfaces including glass and polypropylene stored in liquid nitrogen.
  • They have good print smudge resistance, solvent resistance, and good high and low temperature performance.
  • Specialized – Aggressive adhesive labels are engineered for a wide range of lab ID applications on flat surfaces, including cryogenic storage boxes.
  • Durable – Labels withstand solvents, smudges, liquid nitrogen, freezer applications and a high/low temperature range of -320° to 212°F (-196° to 100°C).
  • Adhesive – Permanent acrylic adhesive for tight bonding to glass, stainless steel and copper, or wrapping around tubes and vials with an 1/8 in. overlap.
Specifications
Specifications
Autoclavable
THT-155-490-3
Permanent Acrylic, Permanent
Matte
Polyester
Cold Surface Label
General identification, Laboratory identification
Roll of 3000 Labels
3000 Pk.
0.6 x 1.625 in. (L x W)
-320°F to +266°F
2.6 mil
17.78 mm
1.62 in.
0.6 in.
RoHS 2002/95/EC
FreezerBondz Thermal Transfer Printer Labels
White
Label
Thermal Transfer
Non-sterile
BBP™81 Label Printer, BRADY IP™ Series Label Printer, BradyPrinter i5100, BradyPrinter i7100, BRADYPRINTER PR Plus Series Label Printer, THT 3 in. Core Printer
1
Rectangle
15.24 x 41.28 mm (L x W)
-196°C to +130 °C
0.065 mm
41.28 x 15 mm
46.36 mm
15 mm