missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Hazwik™ Spill Kits
Chemical class spill kits
£55.24
Product Code | Brand | Capacity (English) | Capacity (Metric) | Includes | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (English) | Capacity (Metric) | Includes | Price | Quantity & Availability | |||
10409894
|
Brady™
813858 |
5 gal. | 15.12 L | (10) 38.1 x 48.26 cm pads, (2) 7.62 cm x 1.22 m socks, nitrile gloves, disposal bag, instruction sheet |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
SKH-CFB Emergency Response Portable Spill Kit
- Reusable clear bag with shoulder strap
- (32) 38.1 x 48.26cm (15 x19 in.) pads
- (5) 7.62cm x 1.22m (3 in. x 4 ft.) SOCs
- 43.18 x 48.26cm (17 x 19 in.) pillow
- Nitrile gloves, goggles, two disposal bags and instruction sheet
- Shipping wt: 11.34kg (25 lbs.)
SKH-PP — Portable Economy Spill Kit
- For small spills of hazardous materials
- (10) 38.1 × 48.26cm (15 × 19 in.) pads
- (2) 7.62cm × 1.22m (3 in. × 4 ft.) SOCs
- Nitrile gloves, disposal bag and instruction sheet
- Shipping wt: 2.26kg (5 lbs.)
Specifications
Portable Spill Containment Kit | |
1 Kit |