missing translation for 'onlineSavingsMsg'
Learn More

Brady™ IDXPERT™ and LABXPERT™ FreezerBondz II Labels: Rectangular

Labels withstand environments from LN2 to autoclave, helping preserve research while maintaining sample traceability

£113.00 - £177.00

Specifications

Description Surface Mount
Adhesive Type Acrylic, Permanent
Color Black, White
Finish Semi-Gloss
Format Label
View More Specs

Products 8
Product Code Brand Quantity Height (English) Width (English) Price Quantity & Availability  
Product Code Brand Quantity Height (English) Width (English) Price Quantity & Availability  
10506360
View Documents
Brady™
105944
225/Pk. 1 in. 0.9 in. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11523834
View Documents
Brady™
105943
1 Ea. 0.5 in. 19 in. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10305213
View Documents
Brady™
105945
225/Pk. 1 in. 1.9 in. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10475563
View Documents
Brady™
105946
275/Pk. 0.37 in. 0.8 in. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10503055
View Documents
Brady™
105947
225/Pk. 0.5 in. 1 in. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11856003
View Documents
Brady™
105948
300/Pk. 0.37 in. 0.37 in. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12839928
View Documents
Brady™
105949
300/Pk. 0.5 in. 0.5 in. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11816013
View Documents
Brady™
105953
250/Pk. 1.25 in. 0.25 in. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Labels withstand environments from LN2 to autoclave, helping preserve research while maintaining sample traceability

  • Made of polyester with permanent acrylic adhesive
  • Can be applied to frozen surfaces including glass and polypropylene stored in liquid nitrogen
  • Provides viewing window for liquid level inside vial
  • Survives exposure to common laboratory environments, such as liquid nitrogen, freezers down to -196°C (-320.8°F), autoclaves, and water baths up to 121°C (249.8°F)
  • Label sizes available for vials, slides, well plates, straws, petri dishes, boxes, and general applications

Order Info

For use with Brady LABXPERT and IDXPERT printers

Specifications

Specifications

Surface Mount
Black, White
Label
Label
iDXPERT™, LabXpert™ Printer
-320.8°F to 249.8°F
0.081 mm
Acrylic, Permanent
Semi-Gloss
Polyester
General and industrial identification, Laboratory identification
1
-196°C to +121°C
Videos
SDS
Documents

Documents

Product Certifications