missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Signs: Stop

STOP ToughStripe™ Floor Signs feature a highly visible, glossy appearance for communicating important safety messages and anti-skid sections to help prevent slips, trips and falls.

Brand:  Brady™ 104511

Product Code. 11746365

  • £128.00 / Each
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • ToughStripe™ Floor Signs communicate important safety and warning messages to your employees in areas with foot traffic
  • Durable ToughStripe™ Anti-Skid Polyester (B-534) includes a rigid, clear overlaminate and is selectively covered by anti-skid tread
  • Highly visible, glossy material resists harsh chemicals and is easy to apply and remove
  • Floor signs alert workers of potential hazards, direct traffic, and enforce protective equipment requirements.
Specifications

Specifications

ANSI A1264.2-2006, OSHA 1910.22
Gloss
1 Ea.
ToughStripe - Pre-printed Floor Signs
431.80 mm
130°F
0.01 in.
Round
English
Floor Marking Sign
B534FS 17 in. DIA RED,BLK/WHT STOP W/SYMBOL
Polyester
Rubber-based
17 in.
For communicating important safety and warning messages to employees
0°F
-18°C to 54°C
0°F to 130 °F
Stop
Black on White
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.