missing translation for 'onlineSavingsMsg'
Learn More

Brady™ TLS 2200™ and TLS PC Link™ General Purpose Labels

TLS-PC Link™ General Purpose Labels are ideal for electronic PCB and component identification, bar code labels, rating plates and solar panel identification.

£31.10 - £88.20

Specifications

Adhesive Type Acrylic, Permanent
Color White
Format Label
Product Type General Purpose Label
For Use With (Equipment) TLS 2200™ or TLS-PC Link™
View More Specs

Products 6
Product Code Brand Quantity Finish Height (Metric) Material Price Quantity & Availability  
Product Code Brand Quantity Finish Height (Metric) Material Price Quantity & Availability  
11767365
View Documents
Brady™
018363
100/Roll Gloss 12 mm Polyester This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10475252
View Documents
Brady™
018503
250/Pk. Matte 38.1 mm Nylon Cloth This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10600473
View Documents
Brady™
018502
500/Pk. Matte 38.1 mm Nylon Cloth This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12867238
View Documents
Brady™
018422
250/Pk. Matte 1.9 cm Vinyl This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12912384
View Documents
Brady™
062267
250/Pk. Matte 2.5 cm Vinyl This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10314982
View Documents
Brady™
018382
500/Pk. Gloss 2.5 cm Polyester This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Labels for the TLS 2200™ and TLS-PC Link™ thermal transfer Portable Printers
  • RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC; UL approved, CSA accepted
  • Glossy white finish with permanent acrylic (B-423) meets requirements of a halogen-free material per DIN VDE 0472 part 815 and has a service temperature of -94 to 230° F (-70 to 110° C)
  • Matte white polyamide coated nylon cloth material (B-499) has a permanent acrylic adhesive with a service temperature of -40 to 193° F (-40 to 90° C) and recommended for indoor use
Specifications

Specifications

Acrylic, Permanent
Label
TLS 2200™ or TLS-PC Link™
White
General Purpose Label
Videos
SDS
Documents

Documents

Product Certifications