Learn More
Brady™ TLS 2200™/TLS PC Link™ Portable Thermal Transfer Printer Labels for Vial Tops
Labels for vial tops
Brand:  Brady™ 062252
 View more versions of this product
Product Code. 10763293
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Labels for vial tops
  • Durable, chemical- and solvent-resistant labels; ink will not smear or fade, and the text stays crisp and readable
  • Printable labels allow more information to fit on any size legend
  • Choice of materials suitable for different environments from liquid nitrogen to autoclaves
Order Info
Labels available in other materials and sizes; contact your Customer Service Representative for ordering information.
Specifications
Specifications
Halogen-free (DIN VDE 0472/parts 815), UL
Acrylic, Permanent
Matte
Nylon Cloth
Laboratory identification, vial, vial top
1
Round
-196°C to +90°C
12.7 mm
9.5 mm
Surface Mount
Yellow
Label
TLS 2200™/TLS-PC Link™ Label
BMP™71, TLS 2200™, TLS-PC Link™
500/Pk.
-320-194°F
0.165 mm
0.375 in.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Brady™ TLS 2200™/TLS PC Link™ Portable Thermal Transfer Printer Labels for Vial Tops > Yellow Matte Nylon Cloth; 0.375 in. dia.