missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ ToughStripe™ Floor Marking Tape
ToughStripe™ Floor Marking Tape (3.5m/100 ft. in length) is a long-lasting, heavy-duty solution for marking floors, aisles, work cells and other designated areas.
£77.45 - £146.00
Specifications
Format | Tape |
---|---|
Material | Polyester |
For Use With (Application) | Used to mark aisles, passageways, storage locations and point out safety equipment and exits |
Quantity | 1 Roll |
Temperature Range (Metric) | -18°C to +54°C |
Product Code | Brand | Color | Width (English) | Width (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Width (English) | Width (Metric) | Price | Quantity & Availability | |||
11791665
|
Brady™
104310 |
Black | 2 in. | 5.08 cm |
£77.45
1 roll |
In Stock
Log in to see stock. |
|||
11791675
|
Brady™
104340 |
Black | 3 in. | 7.62 cm |
£118.00
1 roll |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||
11791685
|
Brady™
104370 |
Black | 4 in. | 10.2 cm |
£146.00
1 roll |
In Stock
Log in to see stock. |
|||
11781675
|
Brady™
104319 |
Black, White | 2 in. | 5.08 cm |
£77.45
1 roll |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||
11781685
|
Brady™
104349 |
Black, White | 3 in. | 7.62 cm |
£118.00
1 roll |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||
11781695
|
Brady™
104379 |
Black, White | 4 in. | 10.2 cm |
£146.00
1 roll |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||
11761675
|
Brady™
104317 |
Black, Yellow | 2 in. | 5.08 cm |
£95.90
1 roll |
In Stock
Log in to see stock. |
|||
11761685
|
Brady™
104347 |
Black, White | 3 in. | 7.6 cm |
£118.00
1 roll |
In Stock
Log in to see stock. |
|||
11761695
|
Brady™
104377 |
Black, Yellow | 4 in. | 10.2 cm |
£146.00
1 roll |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||
11731675
|
Brady™
104314 |
Blue | 2 in. | 5.08 cm |
£84.35
1 roll |
In Stock
Log in to see stock. |
|||
11731685
|
Brady™
104344 |
Blue | 3 in. | 7.62 cm |
£118.00
1 roll |
In Stock
Log in to see stock. |
|||
11731695
|
Brady™
104374 |
Blue | 4 in. | 10.2 cm |
£146.00
1 roll |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||
11741675
|
Brady™
104315 |
Green | 2 in. | 5.08 cm |
£84.40
1 roll |
In Stock
Log in to see stock. |
|||
11741685
|
Brady™
104345 |
Green | 3 in. | 7.62 cm |
£118.00
1 roll |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||
11741695
|
Brady™
104375 |
Green | 4 in. | 10.2 cm |
£146.00
1 roll |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||
11751675
|
Brady™
104316 |
Orange | 2 in. | 5.08 cm |
£84.35
1 roll |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||
11751685
|
Brady™
104346 |
Orange | 3 in. | 7.62 cm |
£118.00
1 roll |
In Stock
Log in to see stock. |
|||
11751695
|
Brady™
104376 |
Orange | 4 in. | 10.2 cm |
£146.00
1 roll |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||
11721675
|
Brady™
104313 |
Red | 2 in. | 5.08 cm |
£84.35
1 roll |
In Stock
Log in to see stock. |
|||
11721685
|
Brady™
104343 |
Red | 3 in. | 7.62 cm |
£118.00
1 roll |
In Stock
Log in to see stock. |
|||
11721695
|
Brady™
104373 |
Red | 4 in. | 10.2 cm |
£146.00
1 roll |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||
11771675
|
Brady™
104318 |
White, Red | 2 in. | 5.08 cm |
£77.45
1 roll |
In Stock
Log in to see stock. |
|||
11771685
|
Brady™
104348 |
White, Red | 3 in. | 7.62 cm |
£118.00
1 roll |
In Stock
Log in to see stock. |
|||
11771695
|
Brady™
104378 |
White, Red | 4 in. | 10.2 cm |
£146.00
1 roll |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||
11701675
|
Brady™
104311 |
White | 2 in. | 5.08 cm |
£80.35
1 roll |
In Stock
Log in to see stock. |
|||
11701685
|
Brady™
104341 |
White | 3 in. | 7.62 cm |
£118.00
1 roll |
In Stock
Log in to see stock. |
|||
11701695
|
Brady™
104371 |
White | 4 in. | 10.2 cm |
£146.00
1 roll |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||
11711675
|
Brady™
104312 |
Yellow | 2 in. | 5.08 cm |
£80.35
1 roll |
In Stock
Log in to see stock. |
|||
11711685
|
Brady™
104342 |
Yellow | 3 in. | 7.62 cm |
£127.00
1 roll |
In Stock
Log in to see stock. |
|||
11711695
|
Brady™
104372 |
Yellow | 4 in. | 10.2 cm |
£146.00
1 roll |
In Stock
Log in to see stock. |
|||
Description
- Visible floor-marking improves safety and organization in almost any industrial environment
- High-gloss surface shines like new paint and withstands forklift traffic without tearing or lifting
- Ideal for marking aisles, passageways and storage locations and identifying equipment and egress paths
- Can be easily applied by one person working alone
Specifications
Tape | |
Used to mark aisles, passageways, storage locations and point out safety equipment and exits | |
-18°C to +54°C | |
30.48 m |
Polyester | |
1 Roll | |
100 ft. |