missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ ToughStripe™ Warning: Forklift Traffic with Pictogram Floor Safety Sign
Use these signs in important places to remind forklift drivers and other warehouse employees to stop or take established paths to avoid potential hazards or accidents.
Brand: Brady™ 104512
Code : NEW
Additional Details : Weight : 0.04990kg
Description
- ToughStripe floor signs feature durable floor marking tape with a high-performance adhesive
- Sign reads WARNING FORKLIFT TRAFFIC (with picto and anti-skid areas)
- Black on yellow
- Diameter: 43.18cm (17 in.)
- B-534 polyester — surface printed polyester with clear polyester overlaminate, anti-skid areas and rubber-based adhesive
- Bright colors and bold text provide high visibility to critical messages
- Designed to withstand harsh industrial environments
Specifications
ANSI A1264.2-2006, OSHA 1910.22 | |
Polyester | |
Permanent Rubber | |
17 in. | |
For communicating important safety and warning messages to employees | |
0°F | |
-18°C to 54°C | |
0°F to 130 °F | |
Warning Forklift Traffic | |
Black on Yellow |
Gloss | |
1 Ea. | |
ToughStripe - Pre-printed Floor Signs | |
431.80 mm | |
130°F | |
0.01 in. | |
Round | |
English | |
Floor Marking Sign |