Learn More
Brady™ BradyJet J2000 Colour Label Printer - UK with Brady Workstation LAB Suite
Product Code. 16364878
Product Code. 16364878
Brand: Brady™ 199967
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The BradyJet J2000 Colour Label Printer is a practical and easy to use, full colour inkjet label printer that can print reliable, high-resolution signs, facility and laboratory sample labels.
The perfect combo. Get the impact of colour—that’s wildly durable—to create a safer, more efficient workplace. Rely on the BradyJet J2000 Colour Label Printer when durable colour matters most.
  • Works great in a multitude of industries, from lab to safety and more
  • Compact design means it can snuggle in to tight spaces
  • On-demand, mid-volume inkjet solution creates photo-quality colour labels in a single pass
  • More colour options than you’d ever need—16 million!
  • Custom identification without the high cost of outsourced printing
  • Includes Brady Workstation Lab Suite to your solution and make label creation even easier
  • Setup is stress free – Contact our Brady Technical Support Team and we will schedule a free phone setup appointment
Specifications
Display Type None
Print Technology Inkjet
Dimensions (L x W) 389 x 264.00 mm
Color Capability Multi-color
Includes This kit contains a J2000 printer, a power cord and AC adapter, a USB cable, a Quick Start Guide, an ink cartridge, a material roll for printhead alignment, a USB stick with drivers and manuals and a Brady Workstation Laboratory Suite
Print Speed 63.50 mm/sec.
Resolution 4800 dpi
Power Supply Details 100/240 V