Learn More
Brady™ BradyJet™ Polyester Tags
Brady™ BradyJet™ Polyester Tags are ideal for a wide range of identification applications and can be use on a variety of laboratory equipment.
Brand:  Brady™ 150012
Additional Details : Weight : 0.45000kg
 View more versions of this product
Product Code. 16130511
  • £123.00 / Each
Estimated Shipment: 06-09-2024
to see stock.
Add to basket
Description
Description
  • Ideal for use in general facility labeling, including accident prevention and confined space tags
  • Compliant with OSHA standard 1910.147 for lockout tagout
  • Compliant with OSHA standard 1910.145 for accident prevention tags
  • Material is a thick polyester tagstock material, with an inkjet receptive vinyl adhered
Specifications
Specifications
Non-adhesive
Matte
Pipe & Valve Marking; Safety & Compliance ID; Visual Workplace Labeling; Warehouse Marking; Lockout Tagout Labeling; GHS & Chemical Labels; Arc Flash Labels
−40 to 140°F
50 ft. x 2.5 in.
White
Polyester
BradyJet™ J2000 Printer
-40°C to +60°C
1.88 in. I.D.