missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bradykinin Acetate Salt

For cellular and molecular biology applications

£187.00 - £319.00

Specifications

Purification Method HPLC
Content And Storage -30°C to -10°C
Format Powder
Molecular Weight (g/mol) 1120.21
Product Type Bradykinin acetate salt
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15473129
View Documents
Thermo Scientific Chemicals
J60345.MC
100 mg
£319.00
100mg
Please to purchase this item. Need a web account? Register with us today!
15463129
View Documents
Thermo Scientific Chemicals
J60345.MB
25 mg
£187.00
25mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Bradykinin Acetate elicits vasoconstriction, a systemic blood pressure reflex, a stimulation of respiration, and contraction of the nictitating membrane. Bradykinin plays an important role in the regulation of fluid and electrolyte balance, smooth muscle contraction, vasodilation and capillary permeability.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

HPLC
Powder
Bradykinin acetate salt
Arg-Pro-Pro-Gly-Phe-Ser-Pro-Phe-Arg-OH
-30°C to -10°C
1120.21
RUO - research use only
Videos
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.