Learn More
BRAND™ EASYCAL™ Professional Calibration Software
Simplifies the tedious task of calibrating liquid handling, glass, and plastic volumetric instruments to ISO 9001 and GLP standards.
Brand: BRAND™ 708440
Code : 02
Additional Details : Weight : 0.11000kg
Description
- Easy-to-follow procedures are outlined step-by-step and all calculations are performed automatically by the software.
- Weighing values may be entered manually into the software or imported directly from the balance via serial port.
- Generates reports to document the calibration testing in accordance to ISO 8655, ISO 4787 etc.
- All test records can be saved in a database for subsequent use and tracking of instrument performance.
- Simplifies data entry
- Organizes all test data
- Includes SOPs
- Clearly arranged test record
Specifications
Calibration Software | |
Calibration of liquid handling to ISO 9001 and GLP standards | |
CD Rom in English, French, German, Portuguese,and Spanish |