missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ BRANDplates hydroGrade™ 96-Well Microplates
Featuring a strongly hydrophilic surface for binding of hydrophilic molecules or for homogeneous assays with hydrophobic molecules that remain in solution
£201.00
Specifications
Binding Property | Hydrophilic |
---|---|
Certifications/Compliance | ANSI, SLAS |
Format | Standard |
Lid | Without Lid |
Array | 8 x 12 |
Product Code | Brand | Color | Well Shape | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Well Shape | Price | Quantity & Availability | ||||
10206813
|
BRAND™
781782 |
Clear | Flat Bottom (F-well) |
£201.00
Pack of 100 |
In Stock
Log in to see stock. |
||||
10012403
|
BRAND™
781780 |
Clear | Round |
£201.00
Pack of 100 |
Estimated Shipment: 19-08-2024
Log in to see stock. |
||||
Description
- Immunoassay surface with strong affinity to hydrophilic molecules, such as glycoproteins and peptides, antibodies with predominantly hydrophilic regions, and nucleic acids
- Blue alphanumeric embossing for easy well identification
- Non-sterile
Order Info
Packaging is 20 stacks of 5 plates each (100 plates). Each stack comes with one lid.
Specifications
Hydrophilic | |
Standard | |
8 x 12 | |
96 | |
Immobilization of hydrophilic molecules |
ANSI, SLAS | |
Without Lid | |
Polystyrene | |
Non-sterile | |
100/Cs. |