Learn More
BRAND™ BRANDplates™ Smooth-Walled Insert Strips
Available with two membranes PC or PET membrane
Brand: BRAND™ 782840
264.10 GBP valid until 2024-12-31
Use promo code "21928" to get your promotional price.
Additional Details : Weight : 0.05600kg
Description
- Sterile
- Smooth-walled
- cellGrade™ plus surface
- Membrane material: PC or PET material for different optical requirements and cell visibility
- Multiple membrane pore sizes for different applications: 0.4μm, 1.0μm, 3.0μm, 8.0μm
- 12 strips/Pk. of four wells each
Applications:
Secretion studies, co-culture, migration studies, chemotaxis assays
Specifications
Polystyrene | |
cellGrade plus | |
3 μm | |
PC |
Sterile | |
Cell Culture Insert Strip | |
12/Pk. |