Learn More
BrandTech™ 96-Well PCR Plate Sealing Mats
Designed specifically for BRAND™ PCR plates. BRAND™ 96-Well PCR Sealing Mats reduce evaporation losses by up to 75 percent in comparison to conventional systems.
Brand: BrandTech™ 781405
Code : LC
Additional Details : Weight : 0.06000kg
Description
Designed specifically for BRAND&;trade; PCR plates.- Reduces evaporation losses by up to 75 percent in comparison to conventional systems
- Highly flexible
- Forms tight seal
- Easily pierced with pipet tips.
Specifications
TPE | |
96 | |
Sealing Mat |
Autoclavable | |
5 | |
96-well PCR Plate |