missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Deep Well Plates

Product Code. p-4363584 Shop All Brand Products
Click to view available options
Volume (Metric) Well:
0.3 mL
0.5 mL
1 mL
1.1 mL
1.2 mL
2.2 mL
Format:
Elevated Wells
Standard
Well Shape:
Round
Round Bottom (U-well)
Square, Conical Bottom (V-well)
Square, Round Bottom (U-well)
Unit Size:
1 pack
Pack of 24
Pack of 32
Pack of 48
Pack of 50
This item is not returnable. View return policy

Product Code. 10680763

Brand: BRAND™ 701346

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Seamlessly integrates into multi-channel systems and automation workflows.

BRAND™ Deep Well Plates are designed for a broad range of applications, including sample storage, high-throughput screening (HTS), cell and tissue culture, serial dilutions, and reagent transfers. Most plates are manufactured from high-quality virgin polypropylene, with one highly transparent polystyrene variant for applications requiring optical clarity.

  • Available in 96- and 384-well formats
  • Manufactured from high-quality virgin polypropylene or polystyrene
  • BIO-CERT™ PCR Quality are free from human DNA, DNase, RNase, and pyrogens
  • Polypropylene plates are autoclavable, chemically resistant, and suitable for storage down to -80°C (-112°F)
  • Polystyrene plate offers excellent transparency and is suitable for storage down to -20°C (-4°F)
  • ANSI/SLAS 1 and 4 compliant for compatibility with automated systems
  • Alphanumeric well labeling and stackable design for efficient organization
  • Notched corners ensure correct orientation in robotic and automated liquid-handling systems
  • Compatible with sealing mats or adhesive films to minimize evaporation and contamination
TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Format Standard
Array 8 x 12
Autoclavable Autoclavable
Color Clear
Product Type 96-well Assay Microplate
Material Polypropylene
Volume (Metric) Well 0.5 mL
No. of Wells 96
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Round Bottom (U-well)
For Use With (Application) Flexible mat covers or adhesive sealing films to reduce sample evaporation and contamination
No. per Pack 48
Profile U-bottom
Quantity 48 Pk.
Mostrar más Mostrar menos
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado