Learn More
BRAND™ Drying Tube
Fits all models of Dispensette™ Digital Buret™ and Titrette™
Brand: BRAND™ 707930
Code : 03
Additional Details : Weight : 0.04000kg
Description
- Without drying agent
- Includes valve block sealing ring
Specifications
Opaque Drying Tube | |
Dispensette™ digital buret |