missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ BRANDplates cellGrade™ 96-Well, cellGrade-Treated, V-Shaped-Bottom Microplate

Color:Clear; Sterility:Sterile; Lid:With Lid

Brand:  BRAND™ 781961

126.17 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.



Code : 03

Additional Details : Weight : 2.71000kg

Product Code. 10002793

  • £166.00 / Pack of 50
In Stock
to see stock.
Add to basket

Description

Description

  • Hydrophilic surface with freely-accessible carboxyl and hydroxyl groups
  • Easily binds serum components to for indirect adhesion of cells
  • Sterilized by β-radiation to ISO 11137 and AAMI guidelines for a SAL of 10-6
  • Meet the requirements of Ph. Eur. and the USP 29
  • DNA-, DNase-, RNase-free
  • Endotoxins are <0.01 EU/mL
  • Orange alphanumeric embossing for easy well identification

Order Info

Packs of 50 individually wrapped plates with lids

Specifications

Specifications

ISO 11137, DIN EN ISO 10993
With Lid
Carboxyl and Hydroxyl Groups
Polystyrene
Sterile
360 μL
Cultivation of adherent cell lines
1/Pk.
Standard
8 x 12
Clear
96
cellGrade
Conical Bottom (V-well)
50/Cs.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers