Learn More
BRAND™ BRANDplates immunoGrade™ 96-Well Microplates
An excellent choice for the majority of standard ELISAs as well as solid phase immunoassays
Brand:  BRAND™ 781724
295.21 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 10477863
  • £335.00 / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Optimized for the immobilization of IgG
  • High binding capacity for molecules with mixed hydrophobic and hydrophilic regions
  • Designed to minimize cross-contamination
  • Blue alphanumeric embossing for easy well identification on standard plates
  • Non-sterile
Order Info
Packaging is 20 stacks of 5 plates each (100 plates). Each stack comes with one lid.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Standard
8 x 12
White
96
330 μL
For the majority of standard ELISAs as well as solid phase immunoassays
Without Lid
High Binding
Polystyrene
Non-sterile
Round
100/Cs.