Learn More
BRAND™ BRANDplates immunoGrade™ 96-Well Microplates
An excellent choice for the majority of standard ELISAs as well as solid phase immunoassays
Brand: BRAND™ 781721
148.91 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Optimized for the immobilization of IgG
- High binding capacity for molecules with mixed hydrophobic and hydrophilic regions
- Designed to minimize cross-contamination
- Blue alphanumeric embossing for easy well identification on standard plates
- Non-sterile
Order Info
Packaging is 20 stacks of 5 plates each (100 plates). Each stack comes with one lid.
Specifications
Standard | |
8 x 12 | |
Clear | |
96 | |
360 μL | |
For the majority of standard ELISAs as well as solid phase immunoassays |
Without Lid | |
High Binding | |
Polystyrene | |
Non-sterile | |
Conical Bottom (V-well) | |
100/Cs. |