missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Self-adhesive Plate Sealing Film
Securely seals multiwell plates. Self-adhesive Plate Sealing Film can be applied directly to plates and removed without the use of expensive equipment, or can be applied with a hard rubber roller to ensure secure and uniform attachment of the film to the plate.
Brand: BRAND™ 701371
50.90 GBP valid until 2024-12-31
Use promo code "21898" to get your promotional price.
This item is not returnable.
View return policy
Description
- Easily apply to plates and remove without the use of expensive equipment.
- Application with a hard rubber roller will ensure secure and uniform attachment of the film to the plate.
- Sealing films are available in different materials especially suited for storage, immunoassay, or cell and tissue culture:
- Polyethylene
- Polyester
- Aluminum
- Rayon
- Polypropylene
- Vinyl
Specifications
Vinyl | |
50 Sheets | |
Storage, fluorescence measurements | |
Sealing Film |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction