missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Self-adhesive Plate Sealing Film

Securely seals multiwell plates. Self-adhesive Plate Sealing Film can be applied directly to plates and removed without the use of expensive equipment, or can be applied with a hard rubber roller to ensure secure and uniform attachment of the film to the plate.

Brand:  BRAND™ 701367

124.36 GBP valid until 2024-12-31
Use promo code "21898" to get your promotional price.


 View more versions of this product

Product Code. 11673119

  • £153.00 / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Easily apply to plates and remove without the use of expensive equipment.
  • Application with a hard rubber roller will ensure secure and uniform attachment of the film to the plate.
  • Sealing films are available in different materials especially suited for storage, immunoassay, or cell and tissue culture:
    • Polyethylene
    • Polyester
    • Aluminum
    • Rayon
    • Polypropylene
    • Vinyl
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DMSO-resistant
100 Sheets
Sealing Film
Polypropylene
Storage
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.