Learn More
BrandTech™ BRAND™ Plastic Cuvettes
BRAND spectrophotometry cuvettes. Mold-cavity matched for the lowest variability in extinction coefficient.
Specifications
Type | Spectrophotometer Cuvette |
---|---|
Disposable | Yes |
For Use With (Application) | Macro and semi-micro sizes to adapt to the needs of most laboratory applications, semi-micro cuvettes require only 1.5mL of sample volume to provide reliable, accurate results |
For Use With (Equipment) | Spectrophotometers and Photometers |
Includes | Arrow mark to indicate direction of transmission and reduce variation |
Description
BRAND spectrophotometry cuvettes. Mold-cavity matched for the lowest variability in extinction coefficient.
- Mold cavity sorted spectrophotometry cuvettes with 2 clear windows
Specifications
Spectrophotometer Cuvette | |
Macro and semi-micro sizes to adapt to the needs of most laboratory applications, semi-micro cuvettes require only 1.5mL of sample volume to provide reliable, accurate results | |
Arrow mark to indicate direction of transmission and reduce variation |
Yes | |
Spectrophotometers and Photometers |