missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ seripettor™ pro Bottletop Dispensers

Product Code. p-4019667 Shop All Brand Products
Click to view available options
Volume (Metric):
0.2 to 2 mL
1 to 10 mL
2.5 to 25 mL
Increments:
0.04 mL
0.2 mL
0.5 mL
Viscosity Rating:
1000mm2/sec.
150mm2/sec.
75 mm2/sec.
Förpackningsstorlek
Each
Denna artikel kan inte returneras. Se returpolicy

Produktkod. 10588153

missing translation for 'mfr': BRAND™ 4720440

Bara null kvar
Lägg i varukorgen
Lägg i varukorgen
Denna artikel kan inte returneras. Se returpolicy

Innehåller

Each seripettor pro bottletop dispenser is supplied with discharge tube, filling tube, spare dispensing catridge, mounting tool, two polypropylene adapters (GL45/33 and GL45/38), operating manual, and one-year warranty.

Designed to handle a wider range of liquids than the regular seripettor™, including certain concentrated acids, polar solvents, and UV-sensitive media.

Key Features:

  • Easily Replaceable Wiping Seal Dispensing Cartridge: Ideal for liquids prone to crystal formation
  • Quick-Change Cartridge: Simplifies maintenance and helps keep lifetime service costs low
  • Corrosion-Resistant Pump Assembly: Features an opaque upper sleeve that shields liquids from UV light during dispensing
  • Telescoping Filling Tube: Adjusts to fit various bottle sizes without the need for cutting
  • Threaded Safety Cap: Attaches and detaches quickly with a simple twist
  • Spring-Loaded Piston: Enables one-handed, semi-automatic operation

Liquids Unsuitable for the seripettorTM Pro:

  • Hydrofluoric Acid and Similar Chemicals: Including those that corrode high-density polyethylene, PTFE, ETFE, FEP, PFA, alumina-ceramic, platinum-iridium, and borosilicate glass
  • Carbon Disulfide
  • Liquids with High Density: Exceeding 2.2 g/cm3
  • Liquids with High Viscosity: Exceeding 300 mm2/5 (for 2 mL instrument), 150 mm2/5 (for 10 mL instrument), or 75 mm2/5 (for 25 mL instrument)
  • Liquids with Vapor Pressure up to 500 mbar

Temperature Guidelines:

  • Instrument and Reagent Temperature: Must be between 59° and 104°F (15° and 407°C)
  • Agar Culture Media: Safe up to 140°F (60°C)

Beställningsinfo

Replaceable piston and cylinder are permanently connected as a cartridge module for safety. If piston, cylinder, or seal are damaged, simply replace cartridge in seconds without tools.

TRUSTED_SUSTAINABILITY

Specifikationer

Accuracy ≤±1.2%
Angle 90°
Product Type Seripettor Pro Bottletop Dispenser
Autoclavable Autoclavable
Color Blue/Red/White
Density Up to 2.2 g/cm3
Channels 1
Material Borosilicate Glass, Ceramic, ETFE, FEP, PFA
Volume (Metric) 1 to 10 mL
Display None
For Use With (Application) Semi-automatic dispensing of many acids, alkaline, and saline solutions
Includes Discharge tube, telescoping filling tube, spare dispensing cartridge, mounting tool, GL45/33mm and GL45/38mm polypropylene adapters, operating manual
Increments 0.2 mL
Precision ≤0.2%
Temperature (English) 59°F to 104°F
Temperature (Metric) 15°C to 40°C
Vapor Pressure Up to 500mbar
Viscosity Rating 150mm2/sec.
Warranty 1 Year
Visa mer Visa mindre
Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.