Learn More
BRAND™ Titrette™ Digital Bottle-Top Burets
GREENER_CHOICE
Provides accuracy and precision within the tolerances of Class A glass burettes. Titrette™ Digital Bottle-top Burets are suitable for most titrants in concentrations up to 1 mol/L. Ideal for general chemistry, wastewater treatment applications, food/beverage analysis, industrial titrations, environmental fieldwork, and more.
£1105.00 - £1810.00
Specifications
Certifications/Compliance ASTM E287, DIN EN ISO 385
Material Glass
View More Specs
Products 6
Product Code Brand Volume (Metric) Includes Price Quantity & Availability  
Product Code Brand Volume (Metric) Includes Price Quantity & Availability  
10703164
GREENER_CHOICE
View Documents Promotion Details
BRAND™
4760151
25 mL (2) AAA Batteries, (2) Amber Inspection Windows, (2) Colored Light Shield Inspection Windows, (3) Bottle Thread Adapters, Operating Manual, Performance Certificate, Telescopic Filling Tube
£1135.00
Each
In Stock
to see stock.
Add to basket
10687425
GREENER_CHOICE
View Documents Promotion Details
BRAND™
4760251
25 mL Performance certificate, telescoping filling tube, 2 AAA batteries, 3 bottle thread adapters, 2 amber inspection windows, operating manual, and RS-232 interface with connector cable and software CD.
£1450.00
Each
Estimated Shipment: 06-05-2025
to see stock.
Add to basket
10703354
GREENER_CHOICE
View Documents
BRAND™
4760161
50 mL (2) AAA Batteries, (2) Amber Inspection Windows, (2) Colored Light Shield Inspection Windows, (3) Bottle Thread Adapters, Operating Manual, Performance Certificate, Telescopic Filling Tube
£1810.00
Each
Estimated Shipment: 28-04-2025
to see stock.
Add to basket
10769165
GREENER_CHOICE
View Documents Promotion Details
BRAND™
4760261
50 mL (2) AAA Batteries, (2) Amber Inspection Windows, (2) Colored Light Shield Inspection Windows, (3) Bottle Thread Adapters, Models With Rs-232 Interface Include Connector Cable and Software Cd, Operating Manua, Performance Certificate, Telescopic Filling Tube
£1460.00
Each
Estimated Shipment: 01-05-2025
to see stock.
Add to basket
15394127
GREENER_CHOICE
View Documents Promotion Details
BRAND™
4760141
10 mL (2) AAA Batteries, (2) Amber Inspection Windows, (2) Colored Light Shield Inspection Windows, (3) Bottle Thread Adapters, Operating Manual, Performance Certificate, Telescopic Filling Tube
£1105.00
Each
In Stock
to see stock.
Add to basket
15604077
GREENER_CHOICE
View Documents Promotion Details
BRAND™
4760241
10 mL (2) AAA Batteries, (2) Amber Inspection Windows, (2) Colored Light Shield Inspection Windows, (3) Bottle Thread Adapters, Models With Rs-232 Interface Include Connector Cable and Software Cd, Operating Manua, Performance Certificate, Telescopic Filling Tube
£1345.00
Each
Estimated Shipment: 28-04-2025
to see stock.
Add to basket
Description
Description
Provides accuracy and precision within the tolerances of Class A glass burettes. BrandTech™ BRAND™ Titrette™ Digital Bottle-top Burets are suitable for most titrants in concentrations up to 1 mol/L. Ideal for general chemistry, wastewater treatment applications, food/beverage analysis, industrial titrations, environmental fieldwork, and more.
The BRAND Titrette provides accuracy and precision within the tolerances of Class A glass burettes. Suitable for most titrants in concentrations up to 1 mol/L, it is ideal for general chemistry, wastewater treatment applications, food/beverage analysis, industrial titrations, environmental fieldwork, and more. The Titrette will enable you to perform routine titrations quickly, accurately, and with ease.
  • Smooth operation for exceptionally sensitive drop-wise titrations
  • Avoids the risk of spills from poured transfers
  • Eliminates meniscus reading errors
  • Switch automatically between filling and titrating
  • Meets tolerances for Class A glass burets per ISO and ASTM standards
  • Optional RS-232 interface available for direct data transmission to a PC
  • Use for a variety of applications including general chemistry, water treatment, food/beverage analysis, industrial titrations, and environmental work in the lab or field
Order Info
For burettes with Calibration Certificate, contact Customer Service. Add DKD at the end of the BRANDTECH part number. No returns accepted.
Specifications
Specifications
ASTM E287, DIN EN ISO 385
Glass
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
BRAND™ Titrette™ Digital Bottle-Top Burets